missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Fura-2 LR/AM, Calbiochem™,

Catalog No. 3449111SET
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3449111SET 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3449111SET Supplier MilliporeSigma™ Supplier No. 3449111SET

Acetoxymethyl (AM) derivative of FURA-PE3 with spectral properties similar to the parent compound

Chemical Identifiers

CAS 172890-84-5
Molecular Formula C55H63N5O29
Molecular Weight (g/mol) 1258.115
InChI Key CGAZNAYDEAJZHQ-UHFFFAOYSA-N
Synonym fura-pe3/am, fura pe 3-am, fura pe-3/am, fura-2 leakres am, fura-2 leakres am , for fluorescence tlc, yl amino-1-benzofuran-2-yl-1,3-oxazole-5-carboxylate, acetyloxymethyl 2-5-2-5-3-4-2-acetyloxymethoxy-2-oxo, acetyloxy methyl 2-2-acetyloxy methoxy-2-oxoethyl 5-2-5-3-4-2-acetyloxy methoxy-2-oxoethyl piperazin-1-yl-3-oxopropyl-2-bis 2-acetyloxy methoxy-2-oxoethyl amino phenoxy ethoxy-2-5-acetyloxy methoxy carbonyl-1,3-oxazol-2-yl-1-benzofuran-6-yl amino acetate, acetyloxy methyl 2-5-2-5-3-4-2-acetyloxy methoxy-2-oxoethyl piperazin-1-yl-3-oxopropyl-2-bis 2-acetyloxy methoxy-2-oxoethyl amino phenoxy ethoxy-6-bis 2-acetyloxy methoxy-2-oxoeth
PubChem CID 16133371
IUPAC Name acetyloxymethyl 2-[5-[2-[5-[3-[4-[2-(acetyloxymethoxy)-2-oxoethyl]piperazin-1-yl]-3-oxopropyl]-2-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]phenoxy]ethoxy]-6-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]-1-benzofuran-2-yl]-1,3-oxazole-5-carboxylate
SMILES CC(=O)OCOC(=O)CN1CCN(CC1)C(=O)CCC2=CC(=C(C=C2)N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C)OCCOC3=C(C=C4C(=C3)C=C(O4)C5=NC=C(O5)C(=O)OCOC(=O)C)N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C

Specifications

Color Yellow
Quantity 1 Set
Solubility Information Soluble in DSMO
Formula Weight 1258.1
Percent Purity ≥95%
Grade HPLC
Physical Form Solid
Chemical Name or Material Fura-2 LR/AM
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above -20°C (-4°F).