CAS RN 17282-00-7
CAS RN 17282-00-7
IUPAC Name:
2-amino-3-bromo-5-methylpyridin-1-ium
Synonyms:
2-amino-3-bromo-5-methylpyridin-1-ium
Molecular Weight (g/mol):
188.05
Molecular Formula:
C6H8BrN2
InChi Key:
NDPKXEWDWTZBDG-UHFFFAOYSA-O
SMILES:
CC1=C[NH+]=C(N)C(Br)=C1
1
–
1
of
1
results
1
2-Amino-3-bromo-5-methylpyridine 98.0+%, TCI America™
CAS: 17282-00-7 Molecular Formula: C6H7BrN2 Molecular Weight (g/mol): 187.04 MDL Number: MFCD00068231 InChI Key: NDPKXEWDWTZBDG-UHFFFAOYSA-N Synonym: 2-amino-3-bromo-5-methylpyridine,2-amino-3-bromo-5-picoline,6-amino-5-bromo-3-picoline,3-bromo-5-methyl-2-pyridinamine,2-amino-5-methyl-3-bromopyridine,2-pyridinamine, 3-bromo-5-methyl,3-bromo-5-methyl-pyridin-2-ylamine,3-bromo-5-methyl-2-pyridylamine,3-bromo-5-methylpyridin-2-ylamine,3-bromo-5-methylpyridin-1-ium-2-amine PubChem CID: 519380 IUPAC Name: 3-bromo-5-methylpyridin-2-amine SMILES: CC1=CC(=C(N=C1)N)Br