missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-bromo-5-methylpyridine 98.0+%, TCI America™

Catalog No. A15555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15555G 5 g
A155525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15555G Supplier TCI America Supplier No. A15555G
Only null left

Chemical Identifiers

CAS 17282-00-7
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
MDL Number MFCD00068231
InChI Key NDPKXEWDWTZBDG-UHFFFAOYSA-N
Synonym 2-amino-3-bromo-5-methylpyridine, 2-amino-3-bromo-5-picoline, 6-amino-5-bromo-3-picoline, 3-bromo-5-methyl-2-pyridinamine, 2-amino-5-methyl-3-bromopyridine, 2-pyridinamine, 3-bromo-5-methyl, 3-bromo-5-methyl-pyridin-2-ylamine, 3-bromo-5-methyl-2-pyridylamine, 3-bromo-5-methylpyridin-2-ylamine, 3-bromo-5-methylpyridin-1-ium-2-amine
PubChem CID 519380
IUPAC Name 3-bromo-5-methylpyridin-2-amine
SMILES CC1=CC(=C(N=C1)N)Br

Specifications

Melting Point 72°C
Quantity 5 g
Formula Weight 187.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-bromo-5-methylpyridine