accessibility menu, dialog, popup

UserName

CAS RN 17135-74-9

BrBr

CAS RN 17135-74-9

IUPAC Name: 1,8-dibromonaphthalene
Synonyms: 1,8-dibromonaphthalene
Molecular Weight (g/mol): 285.97
Molecular Formula: C10H6Br2
InChi Key: DLXBGTIGAIESIG-UHFFFAOYSA-N
SMILES: BrC1=CC=CC2=CC=CC(Br)=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results