missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Dibromonaphthalene 98.0+%, TCI America™
$88.56 - $256.05
Chemical Identifiers
| CAS | 17135-74-9 |
|---|---|
| Molecular Formula | C10H6Br2 |
| Molecular Weight (g/mol) | 285.966 |
| MDL Number | MFCD00183574 |
| InChI Key | DLXBGTIGAIESIG-UHFFFAOYSA-N |
| PubChem CID | 625356 |
| IUPAC Name | 1,8-dibromonaphthalene |
| SMILES | C1=CC2=C(C(=C1)Br)C(=CC=C2)Br |
Chemical Identifiers
| 17135-74-9 | |
| 285.966 | |
| DLXBGTIGAIESIG-UHFFFAOYSA-N | |
| 1,8-dibromonaphthalene |
| C10H6Br2 | |
| MFCD00183574 | |
| 625356 | |
| C1=CC2=C(C(=C1)Br)C(=CC=C2)Br |
Specifications
| 17135-74-9 | |
| 140°C | |
| MFCD00183574 | |
| DLXBGTIGAIESIG-UHFFFAOYSA-N | |
| 1,8-dibromonaphthalene | |
| 625356 | |
| ≥98.0% (GC) | |
| 1,8-Dibromonaphthalene |
| 110°C | |
| C10H6Br2 | |
| 1 g | |
| C1=CC2=C(C(=C1)Br)C(=CC=C2)Br | |
| 285.966 | |
| 285.97 | |
| Crystalline Powder |
Safety and Handling
TSCA : No