accessibility menu, dialog, popup

UserName

H3CCH3CH3

CAS RN 17057-91-9

IUPAC Name: 1,3,8-trimethylnaphthalene
Synonyms: 1,3,8-trimethylnaphthalene
Molecular Weight (g/mol): 170.26
Molecular Formula: C13H14
InChi Key: XYTKCJHHXQVFCK-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C2C(C)=CC=CC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results