missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trimethylnaphthalene 98.0+%, TCI America™

Catalog No. T1712100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1712100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1712100MG Supplier TCI America Supplier No. T1712100MG
Only null left

Chemical Identifiers

CAS 17057-91-9
Molecular Formula C13H14
Molecular Weight (g/mol) 170.255
MDL Number MFCD01321196
InChI Key XYTKCJHHXQVFCK-UHFFFAOYSA-N
PubChem CID 28226
IUPAC Name 1,3,8-trimethylnaphthalene
SMILES CC1=CC=CC2=CC(=CC(=C12)C)C

Specifications

Melting Point 48°C
Color White
Quantity 100 mg
Formula Weight 170.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Trimethylnaphthalene