accessibility menu, dialog, popup

UserName

ONNH

CAS RN 17056-99-4

IUPAC Name: 1,5-dihydroquinoxalin-5-one
Synonyms: 1H-quinoxalin-5-one
Molecular Weight (g/mol): 146.15
Molecular Formula: C8H6N2O
InChi Key: YDXPVVWMFFHSPT-UHFFFAOYSA-N
SMILES: O=C1C=CC=C2NC=CN=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results