missing translation for 'onlineSavingsMsg'
Learn More

5-Quinoxalinol 98.0+%, TCI America™

Catalog No. Q0095200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q0095200MG 200 mg
Q00951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q0095200MG Supplier TCI America Supplier No. Q0095200MG

Chemical Identifiers

CAS 17056-99-4
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.149
MDL Number MFCD00972004
InChI Key YDXPVVWMFFHSPT-UHFFFAOYSA-N
Synonym quinoxalin-5-ol, 5-hydroxyquinoxaline, 5-quinoxalinol, quinoxaline-5-ol, quinoxalin-5 1h-one, ksc172c0d, zuqddqfxsnxeod-uhfffaoysa-n
PubChem CID 12522983
IUPAC Name 1H-quinoxalin-5-one
SMILES C1=CC(=O)C2=NC=CNC2=C1

Specifications

Melting Point 101°C
Color Yellow
Boiling Point 184°C
Quantity 200 mg
Formula Weight 146.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Quinoxalinol