accessibility menu, dialog, popup

UserName

CH3OOSBrN

CAS RN 170235-26-4

IUPAC Name: methyl 2-bromo-1,3-thiazole-4-carboxylate
Synonyms: methyl 2-bromo-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol): 222.06
Molecular Formula: C5H4BrNO2S
InChi Key: YOWKNNKTQWCYNC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CSC(Br)=N1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)