missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromothiazole-4-carboxylate, 96%, Thermo Scientific Chemicals

Catalog No. AAH6354106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6354106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6354106 Supplier Thermo Scientific Chemicals Supplier No. H6354106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 170235-26-4 | Molecular Formula: C5H4BrNO2S | Molecular Weight: 222.056 g/mol

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 170235-26-4
Molecular Formula C5H4BrNO2S
Molecular Weight (g/mol) 222.056
MDL Number MFCD06659908
InChI Key YOWKNNKTQWCYNC-UHFFFAOYSA-N
PubChem CID 2763213
IUPAC Name methyl 2-bromo-1,3-thiazole-4-carboxylate
SMILES COC(=O)C1=CSC(=N1)Br

Specifications

Quantity 5 g
Synonym methyl 2-bromothiazole-4-carboxylate, 2-bromothiazole-4-carboxylic acid methyl ester, 2-bromo-4-methoxycarbonyl-1,3-thiazole, methyl2-bromothiazole-4-carboxylate, methyl-2-bromothiazole-4-carboxylate, 4-thiazolecarboxylic acid, 2-bromo-, methyl ester, 2-bromo-thiazole-4-carboxylic acid methyl ester, 2-bromo-1,3-thiazole-4-carboxylic acid methyl ester, pubchem8920, acmc-1c5qt
Solubility Information Slightly soluble in water.
Formula Weight 222.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material Methyl 2-bromothiazole-4-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only