accessibility menu, dialog, popup

UserName

OHO

CAS RN 1700-30-7

IUPAC Name: [3-(benzyloxy)phenyl]methanol
Synonyms: [3-(benzyloxy)phenyl]methanol
Molecular Weight (g/mol): 214.26
Molecular Formula: C14H14O2
InChi Key: AFKLSWIRJUJWKY-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(OCC2=CC=CC=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results