accessibility menu, dialog, popup

UserName

ONONOO

CAS RN 16968-19-7

IUPAC Name: 1-nitro-2-[2-(2-nitrophenyl)ethyl]benzene
Synonyms: 1-nitro-2-[2-(2-nitrophenyl)ethyl]benzene
Molecular Weight (g/mol): 272.26
Molecular Formula: C14H12N2O4
InChi Key: YBOZRPPSBVIHGJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1CCC1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results