accessibility menu, dialog, popup

UserName

CAS RN 1694-20-8

IUPAC Name: 1-nitro-4-[(1E)-2-phenylethenyl]benzene
Synonyms: trans-4-nitrostilbene 1-nitro-4-styryl-benzene
Molecular Weight (g/mol): 225.25
Molecular Formula: C14H11NO2
InChi Key: ZISCOWXWCHUSMH-VOTSOKGWSA-N
SMILES: [O-][N+](=O)C1=CC=C(\C=C\C2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results