accessibility menu, dialog, popup

UserName

BrON

CAS RN 16883-69-5

IUPAC Name: 1-(2-oxo-2-phenylethyl)pyridin-1-ium bromide
Synonyms: 1-(2-oxo-2-phenylethyl)pyridin-1-ium bromide
Molecular Weight (g/mol): 278.15
Molecular Formula: C13H12BrNO
InChi Key: PXSUMUYPXZEXDT-UHFFFAOYSA-M
SMILES: [Br-].O=C(C[N+]1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)