missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloroanisole, 98%, Thermo Scientific™

Catalog No. p-4089836
Change view
Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398900050 5g Glass bottle
AC398900250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC398900050 Supplier Thermo Scientific Chemicals Supplier No. 398900050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 16817-43-9
Molecular Formula C7H6BrClO
Molecular Weight (g/mol) 221.48
MDL Number MFCD00078659
InChI Key UAMVKOTWSHJOSY-UHFFFAOYSA-N
Synonym 5-bromo-2-chloroanisole, 5-bromo-2-chloroanisol, 4-chloro-3-methoxybromobenzene, 2-chloro-5-bromoanisole, 5-bromo-2-chloro-anisole, benzene, 4-bromo-1-chloro-2-methoxy, 4-bromo-1-chloro-2-methoxy-benzene, zlchem 582, pubchem2632, acmc-209dxq
PubChem CID 2769567
IUPAC Name 4-bromo-1-chloro-2-methoxybenzene
SMILES COC1=C(Cl)C=CC(Br)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 1.6310g/mL
Boiling Point 240°C to 245°C (748.0mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(Cl)OCH3
Quantity 5g
Specific Gravity 1.631
Formula Weight 221.48
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-chloroanisole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only