accessibility menu, dialog, popup

UserName

NHBr

CAS RN 16807-11-7

IUPAC Name: 1-bromo-9H-carbazole
Synonyms: 1-bromo-9H-carbazole
Molecular Weight (g/mol): 246.11
Molecular Formula: C12H8BrN
InChi Key: VCDOOGZTWDOHEB-UHFFFAOYSA-N
SMILES: BrC1=CC=CC2=C1NC1=C2C=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results