missing translation for 'onlineSavingsMsg'
Learn More

1-Bromocarbazole 98.0+%, TCI America™

Catalog No. B4816200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4816200MG 200 mg
B48161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4816200MG Supplier TCI America Supplier No. B4816200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16807-11-7
Molecular Formula C12H8BrN
Molecular Weight (g/mol) 246.107
MDL Number MFCD18450164
InChI Key VCDOOGZTWDOHEB-UHFFFAOYSA-N
PubChem CID 11230335
IUPAC Name 1-bromo-9H-carbazole
SMILES C1=CC=C2C(=C1)C3=C(N2)C(=CC=C3)Br

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 200 mg
Formula Weight 246.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromocarbazole