accessibility menu, dialog, popup

UserName

CAS RN 167218-30-6

HNN

CAS RN 167218-30-6

IUPAC Name: N4,N4,N4'-triphenyl-[1,1'-biphenyl]-4,4'-diamine
Synonyms: N4,N4,N4'-triphenyl-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 412.54
Molecular Formula: C30H24N2
InChi Key: XHPBZHOZZVRDHL-UHFFFAOYSA-N
SMILES: N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results