missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N'-Triphenylbenzidine 97.0+%, TCI America™
$186.41 - $747.49
Chemical Identifiers
| CAS | 167218-30-6 |
|---|---|
| Molecular Formula | C30H24N2 |
| Molecular Weight (g/mol) | 412.536 |
| MDL Number | MFCD17926476 |
| InChI Key | XHPBZHOZZVRDHL-UHFFFAOYSA-N |
| PubChem CID | 22013295 |
| IUPAC Name | N-phenyl-4-[4-(N-phenylanilino)phenyl]aniline |
| SMILES | C1=CC=C(C=C1)NC2=CC=C(C=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC=C5 |
Chemical Identifiers
| 167218-30-6 | |
| 412.536 | |
| XHPBZHOZZVRDHL-UHFFFAOYSA-N | |
| N-phenyl-4-[4-(N-phenylanilino)phenyl]aniline |
| C30H24N2 | |
| MFCD17926476 | |
| 22013295 | |
| C1=CC=C(C=C1)NC2=CC=C(C=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC=C5 |
Specifications
| 167218-30-6 | |
| Yellow | |
| MFCD17926476 | |
| 4-Diphenylamino-4′C-phenylaminobiphenyl | |
| C1=CC=C(C=C1)NC2=CC=C(C=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC=C5 | |
| 412.536 | |
| 412.54 | |
| Crystalline Powder |
| 167°C | |
| C30H24N2 | |
| 1 g | |
| XHPBZHOZZVRDHL-UHFFFAOYSA-N | |
| N-phenyl-4-[4-(N-phenylanilino)phenyl]aniline | |
| 22013295 | |
| ≥97.0% (GC) | |
| N,N,N′-Triphenylbenzidine |
Safety and Handling
TSCA : No