accessibility menu, dialog, popup

UserName

CAS RN 1668-53-7

CH3H3CNNHNH3CCH3OHO

CAS RN 1668-53-7

IUPAC Name: 4-[4,6-bis(2,4-dimethylphenyl)-2,5-dihydro-1,3,5-triazin-2-ylidene]-3-hydroxycyclohexa-2,5-dien-1-one
Synonyms: 4-[4,6-bis(2,4-dimethylphenyl)-5H-1,3,5-triazin-2-ylidene]-3-hydroxycyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 397.48
Molecular Formula: C25H23N3O2
InChi Key: AQFXADRZDKALKG-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(C)=C1)C1=NC(N=C(N1)C1=CC=C(C)C=C1C)=C1C=CC(=O)C=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results