missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2,4-Dihydroxyphenyl)-4,6-bis(2,4-dimethylphenyl)-1,3,5-triazine 98.0+%, TCI America™
$192.00 - $641.24
Chemical Identifiers
| CAS | 1668-53-7 |
|---|---|
| Molecular Formula | C25H23N3O2 |
| Molecular Weight (g/mol) | 397.478 |
| MDL Number | MFCD16251125 |
| InChI Key | AQFXADRZDKALKG-UHFFFAOYSA-N |
| PubChem CID | 5464518 |
| IUPAC Name | 4-[2,6-bis(2,4-dimethylphenyl)-1H-1,3,5-triazin-4-ylidene]-3-hydroxycyclohexa-2,5-dien-1-one |
| SMILES | CC1=CC(=C(C=C1)C2=NC(=C3C=CC(=O)C=C3O)N=C(N2)C4=C(C=C(C=C4)C)C)C |
Chemical Identifiers
| 1668-53-7 | |
| 397.478 | |
| AQFXADRZDKALKG-UHFFFAOYSA-N | |
| 4-[2,6-bis(2,4-dimethylphenyl)-1H-1,3,5-triazin-4-ylidene]-3-hydroxycyclohexa-2,5-dien-1-one |
| C25H23N3O2 | |
| MFCD16251125 | |
| 5464518 | |
| CC1=CC(=C(C=C1)C2=NC(=C3C=CC(=O)C=C3O)N=C(N2)C4=C(C=C(C=C4)C)C)C |
Specifications
| 1668-53-7 | |
| C25H23N3O2 | |
| 5 g | |
| CC1=CC(=C(C=C1)C2=NC(=C3C=CC(=O)C=C3O)N=C(N2)C4=C(C=C(C=C4)C)C)C | |
| 397.478 | |
| 397.48 | |
| Crystalline Powder |
| 205°C | |
| MFCD16251125 | |
| AQFXADRZDKALKG-UHFFFAOYSA-N | |
| 4-[2,6-bis(2,4-dimethylphenyl)-1H-1,3,5-triazin-4-ylidene]-3-hydroxycyclohexa-2,5-dien-1-one | |
| 5464518 | |
| ≥98.0% (HPLC,N) | |
| 2-(2,4-Dihydroxyphenyl)-4,6-bis(2,4-dimethylphenyl)-1,3,5-triazine |
Safety and Handling
TSCA : Yes