accessibility menu, dialog, popup

UserName

CAS RN 16652-71-4

IUPAC Name: benzyl pyrrolidine-2-carboxylate hydrochloridyl
Synonyms: benzyl pyrrolidine-2-carboxylate hydrochloridyl
Molecular Weight (g/mol): 240.71
Molecular Formula: C12H15ClNO2
InChi Key: MRWGIRHDSFQISP-UHFFFAOYNA-N
SMILES: [Cl].O=C(OCC1=CC=CC=C1)C1CCCN1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)