missing translation for 'onlineSavingsMsg'
Learn More

L-Proline Benzyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. P17275G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17275G 5 g
P172725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17275G Supplier TCI America Supplier No. P17275G

Chemical Identifiers

CAS 16652-71-4
Molecular Formula C12H16ClNO2
Molecular Weight (g/mol) 241.715
MDL Number MFCD00039002
InChI Key NEDMOHHWRPHBAL-MERQFXBCSA-N
Synonym l-proline benzyl ester hydrochloride, h-pro-obzl.hcl, l-proline benzyl ester hcl, s-benzyl pyrrolidine-2-carboxylate hydrochloride, benzyl l-prolinate hydrochloride, proline, phenylmethyl ester, hydrochloride, proline benzyl ester hydrochloride, benzyl 2s-pyrrolidine-2-carboxylate hydrochloride, l-proline, phenylmethyl ester, hydrochloride, phenylmethyl 2s pyrrolidine-2-carboxylate, chloride
PubChem CID 2733413
IUPAC Name benzyl (2S)-pyrrolidine-2-carboxylate;hydrochloride
SMILES C1CC(NC1)C(=O)OCC2=CC=CC=C2.Cl

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 5 g
Formula Weight 241.72
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Proline Benzyl Ester Hydrochloride