missing translation for 'onlineSavingsMsg'
Learn More

L-Proline benzyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog No. AAL1561806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1561806 5 g
AAL1561803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1561806 Supplier Thermo Scientific Chemicals Supplier No. L1561806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16652-71-4
Molecular Formula C12H16ClNO2
Molecular Weight (g/mol) 241.715
MDL Number MFCD00039002
InChI Key NEDMOHHWRPHBAL-MERQFXBCSA-N
Synonym l-proline benzyl ester hydrochloride, h-pro-obzl.hcl, l-proline benzyl ester hcl, s-benzyl pyrrolidine-2-carboxylate hydrochloride, benzyl l-prolinate hydrochloride, proline, phenylmethyl ester, hydrochloride, proline benzyl ester hydrochloride, benzyl 2s-pyrrolidine-2-carboxylate hydrochloride, l-proline, phenylmethyl ester, hydrochloride, phenylmethyl 2s pyrrolidine-2-carboxylate, chloride
PubChem CID 2733413
IUPAC Name benzyl (2S)-pyrrolidine-2-carboxylate;hydrochloride
SMILES C1CC(NC1)C(=O)OCC2=CC=CC=C2.Cl

Specifications

Melting Point 146°C to 148°C
Beilstein 3598081
Quantity 5 g
Formula Weight 241.72
Optical Rotation −44° (c=1 in Methanol)
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material L-Proline benzyl ester hydrochloride
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 240-700-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only