CAS RN 16636-51-4
CAS RN 16636-51-4
IUPAC Name:
3-aminocyclohexane-1-carboxylic acid
Synonyms:
3-aminocyclohexane-1-carboxylic acid
Molecular Weight (g/mol):
143.19
Molecular Formula:
C7H13NO2
InChi Key:
CKTUXQBZPWBFDX-UHFFFAOYNA-N
SMILES:
NC1CCCC(C1)C(O)=O
1
–
1
of
1
results
1
cis-3-Aminocyclohexanecarboxylic Acid 96.0+%, TCI America™
CAS: 16636-51-4 Molecular Formula: C7H13NO2 Molecular Weight (g/mol): 143.186 MDL Number: MFCD09753647 InChI Key: CKTUXQBZPWBFDX-RITPCOANSA-N PubChem CID: 3082488 IUPAC Name: (1R,3S)-3-aminocyclohexane-1-carboxylic acid SMILES: C1CC(CC(C1)N)C(=O)O