missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-3-Aminocyclohexanecarboxylic Acid 96.0+%, TCI America™
$361.62 - $1452.10
Chemical Identifiers
| CAS | 16636-51-4 |
|---|---|
| Molecular Formula | C7H13NO2 |
| Molecular Weight (g/mol) | 143.186 |
| MDL Number | MFCD09753647 |
| InChI Key | CKTUXQBZPWBFDX-RITPCOANSA-N |
| PubChem CID | 3082488 |
| IUPAC Name | (1R,3S)-3-aminocyclohexane-1-carboxylic acid |
| SMILES | C1CC(CC(C1)N)C(=O)O |
Chemical Identifiers
| 16636-51-4 | |
| 143.186 | |
| CKTUXQBZPWBFDX-RITPCOANSA-N | |
| (1R,3S)-3-aminocyclohexane-1-carboxylic acid |
| C7H13NO2 | |
| MFCD09753647 | |
| 3082488 | |
| C1CC(CC(C1)N)C(=O)O |
Specifications
| 278°C | |
| White-Yellow | |
| MFCD09753647 | |
| C1CC(CC(C1)N)C(=O)O | |
| 5 g | |
| 3082488 | |
| ≥96.0% (GC,T) | |
| cis-3-Aminocyclohexanecarboxylic Acid |
| 16636-51-4 | |
| C7H13NO2 | |
| CKTUXQBZPWBFDX-RITPCOANSA-N | |
| (1R,3S)-3-aminocyclohexane-1-carboxylic acid | |
| 143.186 | |
| 143.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No