accessibility menu, dialog, popup

UserName

ClSOOS

CAS RN 16629-19-9

IUPAC Name: thiophene-2-sulfonyl chloride
Synonyms: 2-thiophenesulfonyl chloride
Molecular Weight (g/mol): 182.64
Molecular Formula: C4H3ClO2S2
InChi Key: VNNLHYZDXIBHKZ-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=CS1
Boiling Point 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)