missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiophene-2-sulfonyl chloride, 97%
Description
2-Thiophenesulfonyl chloride has been used in the preparation of 2-((trans-2-phenyl cyclopropyl)sulfonyl)thiophene
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Thiophenesulfonyl chloride has been used in the preparation of 2-((trans-2-phenyl cyclopropyl)sulfonyl)thiophene
Solubility
Reacts with water.
Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents, bases.
2-Thiophenesulfonyl chloride has been used in the preparation of 2-((trans-2-phenyl cyclopropyl)sulfonyl)thiophene
Solubility
Reacts with water.
Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents, bases.
Specifications
Specifications
| Melting Point | 29°C to 31°C |
| Boiling Point | 80°C to 81°C (1 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.573 |
| Quantity | 25 g |
| UN Number | UN3261 |
| Beilstein | 123536 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Reacts with water. |
| Formula Weight | 182.65 |
| Show More |
Safety and Handling
| Hazard Category | H314 |
| Hazard Statement | GHS H StatementH314-H318Causes severe skin burns and eye damage.Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| EINECSNumber | 240-677-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only