missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. T21225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T21225G 5 g
T212225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T21225G Supplier TCI America Supplier No. T21225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16629-19-9
Molecular Formula C4H3ClO2S2
Molecular Weight (g/mol) 182.64
MDL Number MFCD00005426
InChI Key VNNLHYZDXIBHKZ-UHFFFAOYSA-N
Synonym 2-thiophenesulfonyl chloride, 2-thienylsulfonyl chloride, thiophene-2-sulphonyl chloride, 2-thiophenesulfonylchloride, 2-thienylsulfonylchloride, 2-thiophene sulfonyl chloride, chloro-2-thienylsulfone, pubchem7737, acmc-209dtw, thiophenesulfonyl chloride
PubChem CID 85518
IUPAC Name thiophene-2-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=CS1

Specifications

Melting Point 33°C
Color White-Yellow
Boiling Point 93°C
Quantity 5 g
UN Number 3261
Formula Weight 182.64
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Thiophenesulfonyl Chloride
TSCA Yes
Recommended Storage Refrigerator