missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxy Naphthol Blue, ACS Reagent, Reagents
Specifications
Specifications
| Chemical Name or Material | Hydroxy Naphthol Blue, ACS Reagent |
| Boiling Point | 1413°C |
| CAS | 165660-27-5 |
| Packaging | Glass Bottle |
| Specific Gravity | 2.17 |
| Molecular Formula | C20H12N2Na2O11S3 |
| Color | Dark Blue |
| Synonym | 1-(2-Naphtholazo-3,6-disulfonic Acid)-2-naphthol-4-sulfonic Acid Disodium Salt |
| Quantity | 125 g |
| Physical Form | Solid |
| Show More |