accessibility menu, dialog, popup

UserName

CAS RN 1655490-79-1

CH3SOOONNN

CAS RN 1655490-79-1

IUPAC Name: {3,13-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-8-ylidene}amino 4-methylbenzene-1-sulfonate
Synonyms: 3,13-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-8-ylideneamino 4-methylbenzenesulfonate
Molecular Weight (g/mol): 351.38
Molecular Formula: C18H13N3O3S
InChi Key: UWCROCJSYGJGEJ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)ON=C1C2=C(N=CC=C2)C2=C1C=CC=N2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results