accessibility menu, dialog, popup

UserName

ONOOOOONOONOONOO

CAS RN 16536-30-4

IUPAC Name: bis(2,4-dinitrophenyl) oxalate
Synonyms: bis(2,4-dinitrophenyl) oxalate
Molecular Weight (g/mol): 422.22
Molecular Formula: C14H6N4O12
InChi Key: CBZOGAWUNMFXFQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OC(=O)C(=O)OC2=CC=C(C=C2[N+]([O-])=O)[N+]([O-])=O)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results