accessibility menu, dialog, popup

UserName

CH

CAS RN 16520-62-0

IUPAC Name: (but-3-yn-1-yl)benzene
Synonyms: but-3-yn-1-ylbenzene
Molecular Weight (g/mol): 130.19
Molecular Formula: C10H10
InChi Key: QDEOKXOYHYUKMS-UHFFFAOYSA-N
SMILES: C#CCCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)