missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1-butyne, 98%

Catalog No. AAL1294614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1294614 25 g
AAL1294606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1294614 Supplier Thermo Scientific Chemicals Supplier No. L1294614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16520-62-0 | C10H10 | 130.19 g/mol

4-Phenyl-1-butyne is used as an intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenyl-1-butyne is used as an intermediate in chemical research.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 16520-62-0
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD00041667
InChI Key QDEOKXOYHYUKMS-UHFFFAOYSA-N
Synonym 4-phenyl-1-butyne, 3-butynylbenzene, but-3-yn-1-ylbenzene, benzene, 3-butynyl, 2-phenylethylacetylene, ccris 7100, phenethylacetylene, 4-phenyl-1-butyn, 3-butynylbenzene #, 4-phenylbut-1-yne
PubChem CID 123360
IUPAC Name but-3-ynylbenzene
SMILES C#CCCC1=CC=CC=C1

Specifications

Density 0.926
Boiling Point 189°C to 190°C
Refractive Index 1.521
Linear Formula C6H5(CH2)2C≡CH
Quantity 25 g
Beilstein 1362221
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 130.19
Percent Purity 98%
Chemical Name or Material 4-Phenyl-1-butyne
Hazard Category H227-H315-H319-H335
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only