missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1-butyne 97.0+%, TCI America™

Catalog No. P03585ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P03585ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P03585ML Supplier TCI America Supplier No. P03585ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16520-62-0
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD00041667
InChI Key QDEOKXOYHYUKMS-UHFFFAOYSA-N
Synonym 4-phenyl-1-butyne, 3-butynylbenzene, but-3-yn-1-ylbenzene, benzene, 3-butynyl, 2-phenylethylacetylene, ccris 7100, phenethylacetylene, 4-phenyl-1-butyn, 3-butynylbenzene #, 4-phenylbut-1-yne
PubChem CID 123360
IUPAC Name but-3-ynylbenzene
SMILES C#CCCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 190°C
Quantity 5 mL
Formula Weight 130.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Phenyl-1-butyne