accessibility menu, dialog, popup

UserName

CAS RN 164462-16-2

H3COOHNOOHOOHNaNaNa

CAS RN 164462-16-2

IUPAC Name: 2-[bis(carboxymethyl)amino]propanoic acid trisodium
Synonyms: 2-[bis(carboxymethyl)amino]propanoic acid trisodium
Molecular Weight (g/mol): 274.14
Molecular Formula: C7H11NNa3O6
InChi Key: SQKOOOSPGWXVRN-UHFFFAOYNA-N
SMILES: [Na].[Na].[Na].CC(N(CC(O)=O)CC(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results