missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trisodium N-(1-Carboxylatoethyl)iminodiacetate Hydrate 95.0+%, TCI America™
$52.19 - $255.38
Chemical Identifiers
| CAS | 164462-16-2 |
|---|---|
| Molecular Formula | C7H11NNa3O6 |
| Molecular Weight (g/mol) | 274.135 |
| InChI Key | SQKOOOSPGWXVRN-UHFFFAOYSA-N |
| Synonym | n,n-bis carboxylatomethyl alanine trisodium salt |
| PubChem CID | 131849341 |
| IUPAC Name | 2-[bis(carboxymethyl)amino]propanoic acid;sodium |
| SMILES | CC(C(=O)O)N(CC(=O)O)CC(=O)O.[Na].[Na].[Na] |
Chemical Identifiers
| 164462-16-2 | |
| 274.135 | |
| n,n-bis carboxylatomethyl alanine trisodium salt | |
| 2-[bis(carboxymethyl)amino]propanoic acid;sodium |
| C7H11NNa3O6 | |
| SQKOOOSPGWXVRN-UHFFFAOYSA-N | |
| 131849341 | |
| CC(C(=O)O)N(CC(=O)O)CC(=O)O.[Na].[Na].[Na] |
Specifications
| 164462-16-2 | |
| C7H11NNa3O6 | |
| n,n-bis carboxylatomethyl alanine trisodium salt | |
| CC(C(=O)O)N(CC(=O)O)CC(=O)O.[Na].[Na].[Na] | |
| 274.135 | |
| 271.11 | |
| Crystalline Powder |
| White-Yellow | |
| 25 g | |
| SQKOOOSPGWXVRN-UHFFFAOYSA-N | |
| 2-[bis(carboxymethyl)amino]propanoic acid;sodium | |
| 131849341 | |
| ≥95.0% (T) | |
| Trisodium N-(1-Carboxylatoethyl)iminodiacetate Hydrate |
Safety and Handling
TSCA : Yes