accessibility menu, dialog, popup

UserName

H3CSOHCHCFe

CAS RN 164297-25-0

IUPAC Name: λ²-iron(2+) 2-(4-methylbenzenesulfinyl)cyclopenta-2,4-dien-1-ide cyclopenta-2,4-dien-1-ide
Synonyms: λ²-iron(2+) 2-(4-methylbenzenesulfinyl)cyclopenta-2,4-dien-1-ide cyclopentadienide
Molecular Weight (g/mol): 324.22
Molecular Formula: C17H16FeOS
InChi Key: QAAXXGKGRWQHLR-UHFFFAOYNA-N
SMILES: [Fe++].[CH-]1C=CC=C1.CC1=CC=C(C=C1)S(=O)C1=CC=C[CH-]1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results