accessibility menu, dialog, popup

UserName

CH3OHH3C

CAS RN 16308-92-2

IUPAC Name: (2,4-dimethylphenyl)methanol
Synonyms: 2,4-dimethylbenzyl alcohol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: QUIMJTKRVOBTQN-UHFFFAOYSA-N
SMILES: CC1=CC=C(CO)C(C)=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)