missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylbenzyl Alcohol 98.0+%, TCI America™

Catalog No. D45195G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D45195G Supplier TCI America Supplier No. D45195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 16308-92-2
Color White-Yellow
Boiling Point 120°C
Molecular Formula C9H12O
Quantity 5 g
MDL Number MFCD00004624
Synonym 2,4-dimethylbenzyl alcohol, 2,4-dimethylphenyl methanol, benzenemethanol, 2,4-dimethyl, 2,4-dimethyl-phenyl-methanol, benzyl alcohol, 2,4-dimethyl, 2,4-dimethylbenzylalcohol, acmc-20aoca, benzenemethanol,2,4-dimethyl, 2,4-dimethylphenyl methanol #
InChI Key QUIMJTKRVOBTQN-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)CO)C
IUPAC Name (2,4-dimethylphenyl)methanol
Molecular Weight (g/mol) 136.194
PubChem CID 27809
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4-Dimethylbenzyl Alcohol
Show More Show Less