accessibility menu, dialog, popup

UserName

CAS RN 162607-20-7

IUPAC Name: (5-methylthiophen-2-yl)boronic acid
Synonyms: 5-methylthiophen-2-ylboronic acid
Molecular Weight (g/mol): 141.98
Molecular Formula: C5H7BO2S
InChi Key: NRIYPIBRPGAWDD-UHFFFAOYSA-N
SMILES: CC1=CC=C(S1)B(O)O
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)