missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiophene-2-boronic acid, 98%

Catalog No. AAB2313806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2313806 5 g
AAB2313803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2313806 Supplier Thermo Scientific Chemicals Supplier No. B2313806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 162607-20-7 | C5H7BO2S | 141.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 162607-20-7
Molecular Formula C5H7BO2S
Molecular Weight (g/mol) 141.98
MDL Number MFCD01318166
InChI Key NRIYPIBRPGAWDD-UHFFFAOYSA-N
Synonym 5-methylthiophene-2-boronic acid, 5-methylthiophen-2-yl boronic acid, 5-methyl-2-thiopheneboronic acid, 5-methyl-2-thienyl boronic acid, 5-methyl-2-thienylboric acid, 5-methylthiophen-2-yl boranediol, 5-methyl-2-thienylboronic acid, 5-methylthiophene-2-boronicacid, 2-methylthiophene-5-boronic acid, 2-methylthienyl-5-boronic acid
PubChem CID 2734374
IUPAC Name (5-methylthiophen-2-yl)boronic acid
SMILES CC1=CC=C(S1)B(O)O

Specifications

Melting Point ∼160°C (decomposition)
Quantity 5 g
Beilstein 7019569
Formula Weight 142.01
Percent Purity 98%
Chemical Name or Material 5-Methylthiophene-2-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only