accessibility menu, dialog, popup

UserName

H2NSSNH2

CAS RN 16214-16-7

IUPAC Name: 2-[(2-aminoethyl)disulfanyl]ethan-1-amine
Synonyms: cystamine
Molecular Weight (g/mol): 152.27
Molecular Formula: C4H12N2S2
InChi Key: APQPRKLAWCIJEK-UHFFFAOYSA-N
SMILES: NCCSSCCN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results