missing translation for 'onlineSavingsMsg'
Learn More

Cystamine Sulfate Hydrate 98.0+%, TCI America™

Catalog No. C1063500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1063500G 500 g
C106325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1063500G Supplier TCI America Supplier No. C1063500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16214-16-7
Molecular Formula C4H14N2O4S3
Molecular Weight (g/mol) 250.346
MDL Number MFCD00060254
InChI Key XVCYODCCYCPADG-UHFFFAOYSA-N
PubChem CID 12998627
IUPAC Name 2-(2-aminoethyldisulfanyl)ethanamine;sulfuric acid
SMILES C(CSSCCN)N.OS(=O)(=O)O

Specifications

Color White
Quantity 500 g
Synonym 2,2′-Diaminodiethyl Disulfide Sulfate, 2,2′-Dithiobis(ethylamine) Sulfate
Formula Weight 250.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cystamine Sulfate Hydrate