accessibility menu, dialog, popup

UserName

HOBOHFF

CAS RN 162101-25-9

IUPAC Name: (2,6-difluorophenyl)boronic acid
Synonyms: 2,6-difluorophenylboronic acid
Molecular Weight (g/mol): 157.91
Molecular Formula: C6H5BF2O2
InChi Key: DBZAICSEFBVFHL-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C=CC=C1F
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results