accessibility menu, dialog, popup

UserName

CAS RN 161950-10-3

IUPAC Name: (4-chloro-3-methylphenyl)boronic acid
Synonyms: 4-chloro-3-methylphenylboronic acid
Molecular Weight (g/mol): 170.4
Molecular Formula: C7H8BClO2
InChi Key: UZDPQDBLCJDUAX-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)