missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methylbenzeneboronic acid, 98%

Catalog No. AAB2317906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2317906 5 g
AAB2317903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2317906 Supplier Thermo Scientific Chemicals Supplier No. B2317906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 161950-10-3 | C7H8BClO2 | 170.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 161950-10-3
Molecular Formula C7H8BClO2
Molecular Weight (g/mol) 170.40
MDL Number MFCD02683104
InChI Key UZDPQDBLCJDUAX-UHFFFAOYSA-N
Synonym 4-chloro-3-methylbenzeneboronic acid, 4-chloro-3-methylphenyl boronic acid, 4-chloro-m-tolueneboronic acid, 4-chloro-m-tolylboronic acid, 4-chloro-3-methyl-phenyl boronic acid, boronic acid, 4-chloro-3-methylphenyl, boronic acid, b-4-chloro-3-methylphenyl, acmc-209xzw, 4-chloro-3-methylphenylboronicacid, 3-methyl-4-chlorophenylboronic acid
PubChem CID 3786632
IUPAC Name (4-chloro-3-methylphenyl)boronic acid
SMILES CC1=C(Cl)C=CC(=C1)B(O)O

Specifications

Melting Point 238°C to 242°C
Quantity 5 g
Formula Weight 170.4
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-methylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only