CAS RN 161485-60-5
CAS RN 161485-60-5
IUPAC Name:
N3,N3,N3',N3'-tetrakis(4-methylphenyl)-[1,1'-biphenyl]-3,3'-diamine
Synonyms:
N3,N3,N3',N3'-tetrakis(4-methylphenyl)-[1,1'-biphenyl]-3,3'-diamine
Molecular Weight (g/mol):
544.74
Molecular Formula:
C40H36N2
InChi Key:
AAVPITVHLAHPPZ-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)N(C1=CC=C(C)C=C1)C1=CC=CC(=C1)C1=CC(=CC=C1)N(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
1
–
1
of
1
results
1
3,3'-Bis[di(p-tolyl)amino]biphenyl 98.0+%, TCI America™
CAS: 161485-60-5 Molecular Formula: C40H36N2 Molecular Weight (g/mol): 544.742 MDL Number: MFCD16036283 InChI Key: AAVPITVHLAHPPZ-UHFFFAOYSA-N PubChem CID: 54123890 IUPAC Name: 3-[3-(4-methyl-N-(4-methylphenyl)anilino)phenyl]-N,N-bis(4-methylphenyl)aniline SMILES: CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=CC(=C3)C4=CC(=CC=C4)N(C5=CC=C(C=C5)C)C6=CC=C(C=C6)C