missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Bis[di(p-tolyl)amino]biphenyl 98.0+%, TCI America™

Catalog No. p-7123806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48741G 1 g
B48745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48741G Supplier TCI America Supplier No. B48741G
Only null left

Chemical Identifiers

CAS 161485-60-5
Molecular Formula C40H36N2
Molecular Weight (g/mol) 544.742
MDL Number MFCD16036283
InChI Key AAVPITVHLAHPPZ-UHFFFAOYSA-N
PubChem CID 54123890
IUPAC Name 3-[3-(4-methyl-N-(4-methylphenyl)anilino)phenyl]-N,N-bis(4-methylphenyl)aniline
SMILES CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=CC(=C3)C4=CC(=CC=C4)N(C5=CC=C(C=C5)C)C6=CC=C(C=C6)C

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 1 g
Formula Weight 544.74
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Bis[di(p-tolyl)amino]biphenyl